Learn More
Medchemexpress LLC Ebvaciclib 25mg | 2185857-97-8 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11417725MG
Encompass_Preferred
Ebvaciclib is a selective, orally bioavailable inhibitor of cyclin-dependent kinases CDK2, CDK4, and CDK6 that induces cell cycle arrest and apoptosis in preclinical tumor models. The compound shows sub-nanomolar to low-nanomolar binding affinity and is supplied as a high-purity research reagent with recommended solvent and formulation guidance for in vitro and in vivo studies.
  • Selective inhibitor of CDK2, CDK4, and CDK6.
  • Orally bioavailable and active in cellular assays.
  • High potency with sub-nanomolar to low-nanomolar affinity.
  • Supplied at high purity for research applications.
  • Soluble in DMSO; example in vivo formulations provided.
Catalog No. 50-003-08161