Learn More
Medchemexpress LLC HY-13615A 5mg , EC-17 (disodium salt) CAS:910661-33-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13615A5MG
Encompass_Preferred
HY-13615A 5mg EC-17 (disodium salt) CAS: 910661-33-5 EC-17 (disodium salt) is a folate receptor alpha (FRα) targeting contrast agent with fluorescent properties in the visible light spectrum. The peak excitation and emission wavelengths of EC-17 are 470/520 nm. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1787