Learn More
Macherey-Nagel HPLC column EC 150/4.6 NUCLEOSIL 100-5 C18
Supplier:  Macherey-Nagel 720120.46
Encompass_Preferred
EC HPLC column - EC 150/4.6 NUCLEOSIL 100-5 C18 - length: 150 mm, ID: 4.6 mm - pack of 1
Catalog No. NC0061034