Learn More
Macherey-Nagel EC HPLC column (analytical), NUCLEOSIL 100-5 C8, 5 um, 250x4.6 mm
Supplier:  Macherey-Nagel 720013.46
Encompass_Preferred
This HPLC column is packed with not endcapped C8 (MOS, monomeric octyl silyl) modification, based on the original NUCLEOSIL silica gel. It is one of the first spherical silicas for HPLC.
Catalog No. NC1811948