Learn More
Macherey-Nagel MACHEREY-NAGEL NUCLEOSIL EC 100-5 C18 HPLC Column, Analytical Grade, 5 µm Particle Size, 200 mm Length
Supplier:  Macherey-Nagel 720006.46
Encompass_Preferred
The MACHEREY-NAGEL NUCLEOSIL EC 100-5 C18 HPLC Column is an advanced analytical column designed for high-performance reversed-phase chromatography. It features a robust stainless steel construction with exceptional pH and temperature stability, ensuring reliable, consistent results for a variety of applications. The fully porous, spherical 5 µm particles provide excellent retention and resolution for effective separation of compounds, making it ideal for routine analytical and preparative tasks.
Catalog No. NC2235339