Learn More
Medchemexpress LLC HY-B0691A 10mg Medchemexpress, Ecabet (sodium) CAS:86408-72-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0691A10MG
Encompass_Preferred
Medchemexpress, HY-B0691A 10mg Ecabet (sodium) CAS:86408-72-2 Ecabet sodium (TA-2711) is currently applied to some clinical gastrointestinal disease by inhibiting the ROS production and improving Helicobacter pylori eradication. Ecabet sodium reduces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0122