Learn More
Medchemexpress LLC eCF309 | 2001571-40-8 | 98.2% | 1 MG

Supplier: Medchemexpress LLC HY1280271MG
eCF309 is a potent and highly selective mTOR inhibitor (IC50 of 15 nM) with higher selectivity over PI3Ks. It is used for the study of breast cancer and prostate cancer, exhibiting highly potent antiproliferative activity against MCF7, MDA-MB-231, and PC3 cells. It also causes G0/G1 cell cycle arrest and inhibits pP70S6K, pS6, and pAKT levels in MCF7 cells.
- Potent and selective mTOR inhibitor
- IC50 of 15 nM
- Higher selectivity over PI3Ks
- Used for breast cancer and prostate cancer studies
- Exhibits antiproliferative activity against MCF7, MDA-MB-231, and PC3 cells
- Causes G0/G1 cell cycle arrest
- Inhibits pP70S6K, pS6, and pAKT levels
- White to off-white solid appearance
- Molecular weight: 383.40
- Chemical formula: C18H21N7O3
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.