Learn More
World Precision Instrument ECG Physiology system
SDP
Supplier:  World Precision Instrument CARDIOPHYS
Encompass
The CardioPhys™ ECG system is a highly versatile tool for measuring ECG of a wide spectra of animals – as small as a zebrafish embryo to animals as large as an alligator.
The CardioPhys™ ECG is a bioamplifier that is designed to amplify the sum of electrical potentials from cardiac muscle, and other extracellular signals. The signal produced from cardiac recordings tracks the depolarization and repolarization of the cardiomyocytes as they contract in succession to pump blood through the heart. The amplifier is powered by two 9 V batteries to minimize electrical noise, which could interfere with or mask the voltage signal from the heart.
Catalog No. 50-253-8910