Learn More
AdipoGen Echinomycin,Chemical. CAS: 512-64-1
SDP
Supplier:  AdipoGen BVT0267M001
Encompass_Preferred
Echinomycin, AdipoGen Life Sciences. Chemical. CAS: 512-64-1. Formula: C51H64N12O12S2. MW: 1101.3. Isolated from Streptomyces echinatus (DSM 40013). Antibiotic. Antitumor compound. Powerful, selective inhibitor of nucleic acid synthesis in vitro. Potent hypoxia-inducible factor 1 (HIF-1) DNA binding activity inhibitor. Apoptosis inducer. Antibacterial, antifungal and antiviral.
Catalog No. 50-464-924