Learn More
ABclonal Technology ECI2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A13072500UL
Encompass_Preferred
This gene encodes a member of the hydratase/isomerase superfamily. The protein encoded is a key mitochondrial enzyme involved in beta-oxidation of unsaturated fatty acids. It catalyzes the transformation of 3-cis and 3-trans-enoyl-CoA esters arising during the stepwise degradation of cis-, mono-, and polyunsaturated fatty acids to the 2-trans-enoyl-CoA intermediates. Alternatively spliced transcript variants have been described.
Catalog No. 50-001-14167