Learn More
New England Biolabs, Inc. EciI – 500 units
SDP
Supplier:  New England Biolabs, Inc. R0590L
Encompass
A restriction endonuclease that recognizes the sequence GGCGGANNNNNNNNN_NN^.
Catalog No. 50-812-301
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product