Learn More
Medchemexpress LLC Eckol 1mg | 88798-74-7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1142031MG
Encompass_Preferred
Eckol is a naturally occurring phlorotannin isolated from brown algae, used in biochemical research as a monoamine oxidase (MAO) inhibitor and for studies of antiviral and antiallergic activity. It is supplied as a white to off-white solid and is provided for research use only (not for human or clinical use).
  • High analytical purity suitable for research applications.
  • Reported hMAO-A and hMAO-B inhibitory activity (IC50 ≈ 7.20 μM and 83.44 μM).
  • Well characterized chemical identity: C18H12O9, MW 372.28 g·mol⁻1.
  • Solid, white to off-white appearance for easy handling.
  • Recommended low-temperature, sealed storage to maintain stability.
  • Intended for in vitro research and biochemical studies.
Catalog No. 50-003-08160