Learn More
LICOR WesternSure ECL Stripping Buffer for Chemiluminescent Westerns (100 mL)
Supplier:  LICOR 92692000
Encompass_Preferred
WesternSure ECL Stripping Buffer is ideal for stripping and reprobing chemiluminescent Western blots. This Western blot stripping buffer. It removes primary and secondary antibodies from PVDF or nitrocellulose membranes, maintains target antigen integrity for efficient reprobing, and it does not require hazardous shipping, unlike many other stripping buffers. Stripping buffers are not recommended if bands need to be quantified. Note: WesternSure ECL Stripping Buffer is not recommended for loading amounts over 30 µg. It will demonstrate decreased performance when used at 2X or 5X concentration and heating the solution will not increase the stripping efficiency.
Catalog No. NC0837896