Learn More
Bio Rad Laboratories ECNO-CLMN CHRMTGRPHY
Supplier:  Bio Rad Laboratories 7372512
Encompass
Column, Chromatography; Bio-Rad; Econo-Column; Glass; Autoclavable; 2.5 x 10cm; 49mL; 2/pk.
Catalog No. 50-901-3430