Learn More
Biolog Inc ECO MICROPLATE
SDP
Supplier:  Biolog Inc 1506
Encompass
This item has to be ordered in multiples of 10 per supplier requirements. ECO MicroPlate - 3 sets of the same 31 carbon sources in a 96 well Biolog MicroPlate. Used for characterization / community analysis).
Catalog No. NC0410462