Learn More
Chemglass Life Sciences ECODryBlock Combo Block for 6mm, 12mm, 13mm, & 25mm Round Bottom Test Tube, 8.3mm O.D x 48.4mm Deep Well, 6 Wells & 13.9mm O.D x 48.4mm Deep Well, 5 Wells & 26.2mm O.D x 48.4mm Deep Well, 3 Wells
SDP
Supplier:  Chemglass Life Sciences CG1991V116
Encompass_Preferred
Provide maximum flexibility, allowing simultaneous heating of different tube formats, ideal for multipurpose laboratories handling diverse workflows.
Catalog No. 50-004-93122