Learn More
Chemglass Life Sciences ECODryBlock System, COD 24, 115-230V
SDP
Supplier:  Chemglass Life Sciences CG1991V24
Encompass_Preferred
ECODryBlock System COD 24 115-230V. Modular digital dry block heater designed for accurate sample heating. Modular digital dry block heater designed for sample preparation incubation DNA denaturation enzymatic reactions culture media tests and COD analysis. ControlPad manages up to two heating Modules with space for up to 2 small aluminum blocks 76 x 51 x 95mm or 1 big aluminum block 152 x 51 x 95mm. Set independent timers and temperatures up to 165°C on each Module. Wide range of interchangeable aluminum blocks to accommodate different types of vials tubes and cuvette. Transparent cover creates warm air jacket that minimizes heat loss and ensures uniform temperature distribution. External probes Pt100 / Pt1000 enable precise monitoring directly within the block or the sample vessel. Possibility to set different temperatures on each Module. 7’’ capacitive touch screen. 3-year warranty. CG-1991-V-24 includes CG-1991-V-204 CG-1991-V-132.
Catalog No. 50-004-93100