Learn More
Medchemexpress LLC Econazole | 27220-47-9 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0885100MG
Encompass_Preferred
Econazole is an orally active imidazole antifungal agent that also functions as a cytochrome P-450 inhibitor and blocks calcium and manganese ion uptake. It demonstrates effectiveness against various fungi and certain Gram-positive bacteria, though it is not significantly active against Gram-negative bacteria. Econazole can inhibit prostaglandin synthesis and may induce liver damage.
  • Orally active imidazole antifungal agent
  • Functions as a cytochrome P-450 inhibitor
  • Blocks calcium and manganese ion uptake
  • Effective against various fungi
  • Effective against some Gram-positive bacteria
  • Can inhibit prostaglandin synthesis
Catalog No. 50-004-08479