Learn More
Medchemexpress LLC Econazole ((±)-Econazol) | 27220-47-9 | 99.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB08851G
Encompass_Preferred
Econazole is an orally active imidazole antifungal agent, a cytochrome P-450 inhibitor, and a blocker of calcium and manganese ion uptake. It is active against various fungi and some Gram-positive bacteria, but not against Gram-negative bacteria. Econazole can inhibit prostaglandin synthesis and can also induce liver damage.
  • Imidazole antifungal agent.
  • Cytochrome P-450 inhibitor.
  • Blocks calcium and manganese ion uptake.
  • Effective against various fungi and some Gram-positive bacteria.
  • Inhibits prostaglandin synthesis.
  • Solid appearance.
  • White to off-white color.
Catalog No. 50-004-08665