Learn More
Medchemexpress LLC Econazole ((±)-Econazol) | 27220-47-9 | MFCD00800993 | 99.93% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB088510MM/1ML
Encompass_Preferred
Econazole is an orally active imidazole antifungal agent. It also acts as a cytochrome P-450 inhibitor and blocks calcium and manganese ion uptake. It demonstrates activity against various fungi and certain Gram-positive bacteria, though it is not significantly active against Gram-negative bacteria. Furthermore, Econazole can suppress prostaglandin synthesis and may induce liver damage.
  • Orally active imidazole antifungal agent
  • Inhibitor of cytochrome P-450
  • Blocks calcium and manganese ion uptake
  • Active against a variety of fungi
  • Active against some Gram-positive bacteria
  • Can inhibit the synthesis of prostaglandins
Catalog No. 50-004-09165