Learn More
Apexbio Technology LLC Econazole nitrate 24169-02-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19375.1
Encompass_Preferred
Econazole nitrate (24169-02-6) is a small-molecule inhibitor targeting the enzyme 14 -demethylase It is designed to disrupt ergosterol synthesis in fungal membrane sterol biosynthesis thereby decreasing membrane integrity altering permeability and attenuating fungal cellular growth Econazole nitrate exerts its biological activity primarily through inhibition of 14 -demethylase In vitro studies demonstrate antifungal activity with reported IC50 values against various fungal isolates typically ranging from approximately 0 05 to 1 g/mL depending on experimental parameters and organism specificity Based on these pharmacological properties econazole nitrate holds research potential in antifungal susceptibility testing exploration of drug resistance mechanisms in fungal strains characterization of dermatopathogenic microorganisms and drug target identification studies
Catalog No. 50-000-88567