Learn More
Medchemexpress LLC Econazole nitrate | 24169-02-6 | 98.0% | 10 G

Supplier: Medchemexpress LLC HYB045310G
Econazole nitrate is an imidazole class antifungal medication that also possesses antibacterial activity.
- Imidazole class antifungal
- Possesses antibacterial activity
- White to off-white solid appearance
- Purity of 98.0%
- Store at 4°C, sealed, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.