Learn More
Medchemexpress LLC Econazole nitrate | 24169-02-6 | MFCD00058160 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB04531G
Encompass_Preferred
Econazole nitrate is an imidazole class antifungal medication that also possesses antibacterial activity. It is clinically used as a topical cream to treat various dermatological infections such as athlete's foot, tinea, pityriasis versicolor, ringworm, and jock itch. Research indicates its efficacy against vaginal candidiasis and good tolerability in toxicity and teratogenicity studies.
  • Imidazole class antifungal
  • Possesses antibacterial activity
  • Treats various skin infections
  • Effective against athlete's foot, tinea, and ringworm
  • Well-tolerated in studies
Catalog No. 50-004-05452