Learn More
Med Vet International Economy Equine Coiled I.V. Tubing Set, J0400LEX, Each
Supplier:  Med Vet International JOR400LEX
Encompass_Preferred
Large IV Set. Affordable at 50% of the cost of standard large bore expandable tubing set. • Large bore tubing for high flow rate • Twin spikes with slide clamp on each • 3 meter long tubing • Roller clamp • Luer lock • Wye injection site. 30cm (12") from distal end • 30cm or 12" spike to burettes • 100cc burette drip chamber with graduations
Catalog No. 50-005-82742