Learn More
MSC Quantum Storage Hopper Shelf Bin, Polypropylene, Yellow, 1 ea
Supplier: MSC 91531053
This heavy-duty shelf storage bin is engineered from high-density polypropylene, providing a robust and versatile solution for organized storage in industrial, metalworking, and MRO environments. The hopper-style bin features a wide front for easy access to stored components and is designed to fit standard 12 in., 18 in., and 24 in. deep shelving units or racks.
- Durable high-density polypropylene construction with reinforced edges
- Built-in rear hang-lock for added stability on compatible shelving
- Molded-in label holder for efficient inventory organization
- Nestable design for space-saving storage when not in use
- Maximum load capacity of 75 lb.
- Operating temperature range up to 250°F
- Seven divider slots accommodate optional dividers for internal compartment configuration