Learn More
World Precision Instrument Economy Tweezers #3C, 11cm, 0.4x0.45mm Tips
SDP
Supplier:  World Precision Instrument 501977
Encompass
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.
Catalog No. 50-252-1368