Learn More
Electron Microscopy Sciences EMS Premium Economy Tweezers Style 7, 120 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7832682
Encompass_Preferred
The tweezers are made from 316 Stainless Steel and are anti-acid/non-corrosive and anti-magnetic. These inexpensive tweezers may be beneficial in non-critical applications.
Tips: 120 mm
OAL: Fine, curved
Catalog No. 50-193-0948