Learn More
Research Products International Corp Econo-Safe Economical Biodegradable Counting Cocktail, 5 Gallons
SDP
Supplier:  Research Products International Corp 111176
Encompass_Preferred
Econo-Safe™ meets the demand for a general purpose scintillation cocktail with a value price. Econo-Safe has a sample holding capacity of up to 20% aqueous volume within a clear, stable region. Ideal for all routine counting applications of aqueous and non-aqueous samples including radiation wipe tests. All components in Econo-Safe are biodegradable and non-hazardous.
  • Low odor
  • High flash point (>300°F)
  • Low viscosity formula
  • Does not require dilution prior to use
Catalog No. 50-212-985