Learn More
Medchemexpress LLC Ecotin E. coli His 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP790835UG
Encompass_Preferred
Ecotin a bacterial protein inhibitor regulates serine proteases like trypsin and subtilisin Ecotin s unique surface loop forms a stable complex influencing enzymatic activity Ecotin Protein E coli (His) is the recombinant E coli-derived Ecotin protein expressed by E coli with C-His labeled tag
Catalog No. 50-001-94601