Learn More
Medchemexpress LLC Ecteinascidin 770 | 114899-80-8 | 99.8% | 770.85 | C40H42N4O10S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101191100MG
Encompass_Preferred
Ecteinascidin 770 (ET-770) is a 1,2,3,4-tetrahydroisoquinoline alkaloid showing potent in vitro anticancer activity. It inhibits multiple human cancer cell lines at low-nanomolar concentrations (for example, U373MG IC50 = 4.83 nM) and can sensitize certain lung cancer cells to anoikis through p53-mediated modulation of MCL1 and BAX. The compound is supplied as a high-purity solid for research use.
Catalog No. 50-002-13170