Learn More
Medchemexpress LLC Ecteinascidin 770 | 114899-80-8 | 99.8% | 770.85 g·mol⁻¹ | C40H42N4O10S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10119125MG
Encompass_Preferred
Ecteinascidin 770 (ET-770) is a marine-derived tetrahydroisoquinoline alkaloid with potent cytotoxic and anticancer activity, commonly used as a research reagent for in vitro pharmacology and cell biology studies. The compound is supplied as a solid (white to yellow) and is typically prepared as DMSO stock solutions for cellular assays.
  • Potent cytotoxic activity against multiple cancer cell lines (low nanomolar IC50s).
  • High reported chemical purity suitable for research applications.
  • Available in multiple small-scale quantities for assay development and screening.
  • Stable when stored cold and protected from light; specific storage recommendations for solids and solutions.
  • Provided with chemical identifiers and safety documentation for laboratory handling.
Catalog No. 50-002-12079