Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ecteinascidin 770 | 114899-80-8 | 99.8% | 770.85 g/mol | C40H42N4O10S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1011915MG

Encompass_Preferred

Ecteinascidin 770 (ET-770) is a tetrahydroisoquinoline alkaloid research compound with potent anticancer activity. It is supplied as a solid (white to yellow) and as a DMSO solution for laboratory use; intended for in vitro research and should be handled according to supplier documentation and SDS guidance.

  • Potent anticancer activity; IC50 4.83 nM against U373MG cells.
  • High purity typically around 99.8%.
  • Molecular formula C40H42N4O10S and molecular weight 770.85 g/mol.
  • Available in small-scale solid pack sizes and as a 10 mM solution in DMSO.
  • Solid appearance: white to yellow powder; soluble in DMSO for assays.

Catalog No. 50-002-12685


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.