Learn More
Medchemexpress LLC Edar protein, human (HEK293, His) | >95.0% | 50 UG
SDP
Supplier:  Medchemexpress LLC HYP7526250UG
Encompass_Preferred
Recombinant human EDAR protein expressed in HEK293 cells with a C-terminal His tag. The protein interacts with EDA-A1 and activates NF-κB and JNK signaling, making it suitable for receptor-ligand interaction studies, signaling assays, and biochemical characterization. Supplied lyophilized with reconstitution and storage guidance.
  • Recombinant human protein produced in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • High purity: >95% as determined by reducing SDS-PAGE.
  • Lyophilized from PBS, pH 7.4, for enhanced stability.
  • Apparent molecular weight ~25-35 kDa due to glycosylation.
  • Suitable for receptor binding, signaling, and biochemical assays.
  • Store lyophilized at -20°C; after reconstitution stable short-term at 4°C; freeze aliquots for long-term storage.
Catalog No. 50-002-92880