Learn More
Selleck Chemical LLC Edaravone-50mg
SDP
Supplier:  Selleck Chemical LLC S132650MG
Encompass_Preferred
Edaravone (MCI-186) is a novel potent free radical scavenger that has been clinically used to reduce the neuronal damage following ischemic stroke.
Catalog No. 50-239-0029