Learn More
Medchemexpress LLC Edaravone | 89-25-8 | 99.9% | C10H10N2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB009910MM/1ML
Encompass_Preferred
Edaravone is a potent, novel free radical scavenger. It inhibits MMP-9-related brain hemorrhage, demonstrates neuroprotective effects by ameliorating neuronal damage in brain ischemia, and offers preventative and therapeutic benefits against glutamate-induced toxicity.
  • Inhibits MMP-9-related brain hemorrhage.
  • Shows neuroprotective effects in brain ischemia.
  • Increases beneficial eNOS.
  • Reduces post-reperfusion brain edema and hemorrhagic events.
  • Significantly decreases infarct volume and postischemic hemorrhage.
  • Decreases astrocyte activity and apoptotic cells.
  • Protects against glutamate-induced neurotoxicity.
  • Associated with the PI3K/Akt pathway and Bcl-2 protein family.
Catalog No. 50-004-32826