Learn More
Medchemexpress LLC Edaravone | 89-25-8 | MFCD00003138 | >99% | C10H10N2O | 10 G
SDP
Supplier:  Medchemexpress LLC HYB009910G
Encompass_Preferred
Edaravone is a potent free radical scavenger, noted for its ability to inhibit MMP-9-related brain hemorrhage. It is also used to ameliorate symptoms of amyotrophic lateral sclerosis (ALS). For storage, keep at -20°C in a sealed container, away from moisture.
  • Attenuates ferroptosis and reduces lipid peroxidation in cells.
  • Increases glutathione levels and inhibits xCT protein level decrease.
  • Enhances antioxidant enzyme activities and upregulates glutathione peroxidase.
  • Improves motor function and survival in ALS mouse models.
  • Suppresses neuroinflammation and microglial activation.
  • Improves mitochondrial integrity and function.
  • Reduces early brain injury and brain edema in intracerebral hemorrhage models.
Catalog No. 50-004-32961