Learn More
COMPLETE GREEN COMPANY EDDHA ACID FREE 4G
Supplier:  COMPLETE GREEN COMPANY EDDHA4G
Encompass
Ethylenediamine-di (o-hydroxyphenylacetic acid), 1170-02-1, C18O6N2H20 - function - used to sequester iron in cell tissue culture
Catalog No. NC0479304