Learn More
Aviva Systems Biology Corp EDDM3B Antibody - middle region (ARP53756_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53756_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EDDM3B. It was validated on Western Blot. . Alternate names: EP3B, FAM12B, HE3-BETA, HE3BApplicationWB. Species reactivity: Human
Catalog No. 50-285-879