Learn More
Medchemexpress LLC Edelfosine | 77286-66-9 | MFCD00135190 | 99.9% | 523.73 g·mol⁻¹ | C27H58NO6P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1086105MG
Encompass_Preferred
Edelfosine ((R)-ET-18-OCH3) is an ether lipid analog used in research for its antiviral and antineoplastic properties. Supplied for laboratory research use only, it is provided as a high-purity reagent for in vitro studies of apoptosis, lipid-mediated signaling, and related biochemical assays.
  • High purity (99.9%) for reliable experimental results.
  • Ether lipid analog with demonstrated anti-HIV and antineoplastic activity.
  • Molecular weight 523.73 g·mol⁻¹ and chemical formula C27H58NO6P for reference.
  • Available in small mg-scale pack sizes suitable for laboratory research.
  • Datasheet and safety data sheet available to support handling and storage.
Catalog No. 50-002-64138