Learn More
Medchemexpress LLC Edetate calcium disodium dihydrate | 6766-87-6 | MFCD00149454 | 98.0% | 410.30 g/mol | C10H16CaN2Na2O10 | 1g
SDP
Supplier:  Medchemexpress LLC HYW7498671G
Encompass_Preferred
Ca -EDTA disodium dihydrate (Calcium disodium EDTA dihydrate) is an orally active metal chelating reagent exhibits bactericidal activities against periodontal pathogens Aggregatibacter actinomycetemcomitans Prevotella intermedia and Porphyromonas gingivalis[1] Ca -EDTA disodium dihydrate is effective chelating antidotes for lead-and cadmium poisoning[2][3]
Catalog No. 50-002-69319