Learn More
Eurofins Discoverx PathHunter EDG1 Total Internalization HEK 293 Cell Line, 2 vials. (93-0784C1)
Supplier:  Eurofins Discoverx 930784C1
Encompass
The PathHunter EDG1 Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of EDG1 via detection of localization to the early endosome. The included cell line overexpresses EA-tagged EDG1 and a ProLink-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the EDG1 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1307
Only null left
Add to Cart
Request a Quote