Learn More
Medchemexpress LLC Edivoxetine hydrochloride | 1194374-05-4 | C18H27ClFNO4 | 100 MG

Supplier: Medchemexpress LLC HY15413A100MG
Edivoxetine hydrochloride (LY 2216684) is a potent and selective norepinephrine reuptake inhibitor. It is used for the study of major depressive disorder (MDD) and attention-deficit/hyperactivity disorder (ADHD).
- Potent and selective norepinephrine reuptake inhibitor
- Aids in the study of major depressive disorder (MDD)
- Aids in the study of attention-deficit/hyperactivity disorder (ADHD)
- Purity of 99.85%
- Appears as a white to off-white solid
- Soluble in DMSO at 125 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.