Learn More
Medchemexpress LLC Edivoxetine hydrochloride | 1194374-05-4 | C18H27ClFNO4 | 50 MG

Supplier: Medchemexpress LLC HY15413A50MG
Edivoxetine (LY 2216684) hydrochloride is a potent and selective norepinephrine reuptake inhibitor. It can be used for the study of major depressive disorder (MDD) and attention-deficit/hyperactivity disorder (ADHD). It is for research use only and not sold to patients.
- Potent and selective norepinephrine reuptake inhibitor
- For research use only
- Molecular weight: 375.86
- Appearance: white to off-white solid
- Purity: 99.85%
- Store at -20°C in sealed storage, away from moisture
- In solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.