Learn More
Medchemexpress LLC Edivoxetine hydrochloride | 1194374-05-4 | MFCD27987928 | 99.9% | 375.86 g·mol-1 | C18H27ClFNO4 | 10 MG

Supplier: Medchemexpress LLC HY15413A10MG
Edivoxetine hydrochloride is the hydrochloride salt of edivoxetine, a potent and selective norepinephrine reuptake inhibitor used in research on major depressive disorder and related indications. It is supplied as a high-purity solid for laboratory and pharmacological studies.
- Potent, selective norepinephrine reuptake inhibitor for pharmacology studies.
- High purity (≈99.9%) suitable for analytical and in vitro applications.
- Solid powder form, easy to weigh and dissolve for assays.
- Molecular formula C18H27ClFNO4; molecular weight 375.86 g·mol-1.
- Available in small pack sizes including 10 mg for scarce-sample work.
- Store at -20°C, sealed and protected from moisture.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.