Learn More
TARGETMOL CHEMICALS INC EDO-S101 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T351610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EDO-S101 (Tinostamustine) is a pan HDAC inhibitor with IC50 values of 9 9 and 25 nM for HDAC1 HDAC2 and HDAC3 respectively. purity: 98%
Catalog No. 50-257-6925