Learn More
Medchemexpress LLC Edoxaban | 480449-70-5 | MFCD13195544 | 99.9% | 548.06 g·mol⁻¹ | C24H30ClN7O4S | 25 MG

Supplier: Medchemexpress LLC HY10264R25MG
Edoxaban (Standard) is an analytical reference standard of the oral direct Factor Xa inhibitor edoxaban, intended for research, method development, and quality control in laboratory settings.
- Analytical reference standard for method development and quality control.
- High purity: 99.9% (HPLC) as shown in certificate of analysis.
- Chemical formula: C24H30ClN7O4S; molecular weight: 548.06 g·mol⁻¹.
- CAS number: 480449-70-5.
- Supplied in small, laboratory-scale quantities suitable for analytical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.