Learn More
Medchemexpress LLC Edrophonium chloride | 116-38-1 | 99.39% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0882100MG
Encompass_Preferred
Edrophonium chloride is a short-acting acetylcholinesterase (AChE) inhibitor. It can be utilized to detect early digitalis toxicity and in the study of myasthenia gravis.
  • Exhibits Ki values of 0.2, 0.2, and 0.4 μM for AChE in human erythrocytes, purified calf forebrain, and purified octopus brain, respectively.
  • Shows IC50 values of 0.2, 0.05, and 0.5 μM for AChE in human erythrocytes, purified calf forebrain, and purified octopus brain, respectively.
  • Inhibits DMPP-induced currents in Xenopus laevis oocytes expressing mouse muscle nicotinic acetylcholine receptor (nAChR) with an IC50 of 82.1 μM.
Catalog No. 50-004-09163