Learn More
Apexbio Technology LLC Edrophonium (chloride),250mg CAS# 116-38-1
SDP
Supplier:  Apexbio Technology LLC C4140250
Encompass_Preferred
Edrophonium is a competitive inhibitor of acetylcholinesterase (AChE) [1]. AChE is an extrinsic membrane-hound enzyme that functions in the central and peripheral nervous systems. AChE rapidly terminates the ACh receptor-mediated signal transmission by hydrolyzing Ach. Inhibition of AChE results in accumulation of ACh in the synaptic cleft and leads to impeded neurotransmission [2].In vitro: Edrophonium inhibited AChE activity in human red blood cells, purified calf forebrain, and octopus brain with Ki values of 0.2, 0.2, and 0.4 μM, respectively. The IC50s were 0.2, 0.05, and 0.5 μM, respectively [1]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0710