Learn More
Selleck Chemical LLC Edrophonium chloride
SDP
Supplier:  Selleck Chemical LLC S59005MG
Encompass_Preferred
Edrophonium chloride is the chloride salt of edrophonium which is a rapid-onset short-acting readily reversible inhibitor of acetylcholinesterase
Catalog No. 50-000-03733