Learn More
Kd Medical Inc EDTA 0.5M PH 8.0
SDP
Supplier:  Kd Medical Inc RGF3130
Encompass_Preferred
EDTA is used extensively in molecular biology to minimize metal ion impurities in reaction buffers. Ideal for DNA work. One 1 Liter bottle.
Catalog No. 50-101-9076